Please login to your online account to display your discounted pricing

Iodine monochloride solution, Macron Fine Chemicals™


macron

Quantity 1L

Manufacturer: avantor performance  H18005

 View more versions of this product

Catalog No. MKH1801

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 162.35
Physical Form Liquid
Color Red-brown
Chemical Name or Material Iodine monochloride solution
Molecular Formula ICl
Synonym Chloroiodane
Quantity 1L

Specifications

Chemical Name or Material Iodine monochloride solution
Physical Form Liquid
Molecular Formula ICl
Formula Weight 162.35
Synonym Chloroiodane