missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine monochloride solution, Macron Fine Chemicals™

macron

Chemical Identifiers

Physical Form Liquid
Molecular Formula ICl
Formula Weight 162.35
Synonym Chloroiodane
Chemical Name or Material Iodine Monochloride Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MKH1801 Avantor Performance
H18005
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKH1804 Avantor Performance
H18007
4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

Liquid
162.35
Iodine Monochloride Solution
ICl
Chloroiodane

Specifications

162.35
Red-brown
Chloroiodane
Iodine monochloride solution
Liquid
ICl
1L