Please login to your online account to display your discounted pricing

Iodine monochloride, ACS reagent, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412361000

 View more versions of this product

Catalog No. AC412361000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Fuming Liquid
Formula Weight 162.36
Color Deep reddish-brown
Quantity 100g
Merck Index 15, 5060
Assay 1.0 to 1.2 (I/Cl ratio of the WIJS solution prepared from the reagent)
EINECS Number 232-236-7
TSCA TSCA
Density 3.2000g/mL
RTECS Number NN1650000
Fieser 01,502; 08,261; 10,212; 11,268
Chemical Name or Material Iodine monochloride
Packaging Glass bottle
CAS 7790-99-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Insoluble Matter 0.005% max.
Molecular Formula ClI
Linear Formula ICl
MDL Number MFCD00011354

Specifications

Chemical Name or Material Iodine monochloride
CAS 7790-99-0
Purity Grade ACS Reagent Grade
Assay 1.0 to 1.2 (I/Cl ratio of the WIJS solution prepared from the reagent)
Molecular Formula ClI
Linear Formula ICl
Formula Weight 162.36
MDL Number MFCD00011354
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents