Learn More
Iodine monochloride, ACS, Thermo Scientific Chemicals
Moisture and Light Sensitive
Supplier: Thermo Scientific Chemicals 04434814
Description
Iodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
96°C to 98°C | |
25°C to 27°C | |
7790-99-0 | |
Crystalline, Liquid | |
ClI | |
3902972 | |
Soluble in water,alcohol,ether,carbon disulfide. | |
ClI | |
162.35 | |
162.36 | |
Light and moisture sensitive | |
3.182 |
Iodine monochloride | |
For analysis ACS | |
Black-Red | |
25 g | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
ACS | |
Red-black crystalline/liquid |
Chemical Identifiers
7790-99-0 | |
162.35 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 | |
ClI |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
iodochlorane |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H300-H311-H314-H335-H373
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 232-236-7
RTECSNumber : NN1650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only