Learn More
Iodine monochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196961000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
97.0°C | |
26.0°C | |
7790-99-0 | |
Fuming Liquid | |
97.5% min. (Iodometry) | |
ICl | |
01,502; 08,261; 10,212; 11,268 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
98% | |
3.2 |
Iodine monochloride | |
98% | |
Red to Brown | |
100 g | |
ClI | |
MFCD00011354 | |
15, 5060 | |
Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2 | |
ClI | |
162.35 | |
162.36 | |
Glass bottle | |
3.2000g/mL |
Chemical Identifiers
7790-99-0 | |
162.35 | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
24640 | |
ClI |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
iodochlorane |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 232-236-7
RTECSNumber : NN1650000
TSCA : TSCA
RUO – Research Use Only