missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iodine monochloride, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 428941000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iodine monochloride | |
Liquid | |
ClI | |
MFCD00011354 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
0.95 to 1.05M | |
1.42 |
7790-99-0,75-09-2 | |
100 mL | |
ICl | |
15, 5060 | |
Solubility in water: insoluble | |
ClI | |
162.35 | |
162.35 | |
AcroSeal™ Glass Bottle | |
1.4200g/mL |
RUO – Research Use Only