Learn More
Iodine monobromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 208840500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
116.0°C | |
42.0°C to 50.0°C | |
Black-Brown to Gray | |
50 g | |
BrI | |
MFCD00011353 | |
15, 5059 | |
Solubility in water: soluble. Other solubilities: solubel in alcohol, ether, carbon disulfide and, glacial acetic acid | |
BrI | |
206.81 | |
206.81 | |
Glass bottle | |
4.4100g/mL |
Iodine monobromide | |
7789-33-5 | |
Crystalline Mass, Crystals or Chunks | |
97.5% min. (Iodometry) | |
IBr | |
01,501; 02,224 | |
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution | |
CBEQRNSPHCCXSH-UHFFFAOYSA-N | |
iodobromane | |
82238 | |
98% | |
4.41 |
Chemical Identifiers
7789-33-5 | |
206.81 | |
CBEQRNSPHCCXSH-UHFFFAOYSA-N | |
82238 | |
BrI |
BrI | |
MFCD00011353 | |
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution | |
iodobromane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 232-159-9
RUO – Research Use Only