Please login to your online account to display your discounted pricing

Iodine, Lugol's, Certified, 5.0% (w/v) ±0.5%, LabChem™

$30.91 - $30.91

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 85
CAS 7681-11-0
CAS Max % 10
CAS 7553-56-2
CAS Max % 5
Name Note Lugol's
Purity Grade Certified Grade
Linear Formula I2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156757

 
lab chem
LC156757
Amber Glass 125mL Each for $30.91
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 5.0% ± 0.5% (w/v)
Formula Weight 253.809
CAS 7732-18-5
Chemical Name or Material Iodine
CAS Max % 5
Packaging Amber Glass
Name Note Lugol's
Physical Form Liquid
Linear Formula I2
Storage Information Room Temperature
Solubility Information Soluble in water
Health Hazard 1 Warning
Color Dark red-brown
Quantity 125mL
CAS Max % 85
Health Hazard 2 GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.
CAS 7681-11-0
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Iodine vapor; Potassium oxide
CAS Max % 10
Traceability to NIST Traceable to NIST
CAS 7553-56-2
Identification Passes Test