missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine (Iodine-Iodide), 0.100 Normal, Ricca Chemical

$397.97 - $1,783.03

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.809
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
40-003-2CS
View Documents
Ricca Chemical Company
RIR40000006X1C
6L Amber glass
Each for $397.97
 
40-001-CS
View Documents
Ricca Chemical Company
RIR40000004X4C
16L Amber glass
Each for $1,783.03
 
Specifications

Chemical Identifiers

7553-56-2
253.809
diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
CHEBI:17606
II
I2
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
molecular iodine

Specifications

7553-56-2,7681-11-0
I2
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
Liquid
Amber glass
Colorless
Iodine (Iodine-Iodide), 0.100 N (N/10)
0.1000 ± 0.0002 N at 20°C
diiodine, eranol, iode, iodine, iodine crystals, iodine solution, iodine sublimed, iodio, tincture iodine, vistarin
II
253.809
CHEBI:17606
Characteristic iodine odor
Nonaqueous Solution
6L
SDS