Please login to your online account to display your discounted pricing

Iodine, Gram Solution


Quantity 100mL
Packaging 0.25N Aqueous Solution

Manufacturer: fisher scientific  IX1802

 View more versions of this product

Catalog No. S71310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100mL
Color Amber
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Packaging 0.25N Aqueous Solution
ChemAlert Storage Symbol Gray
Chemical Name or Material Iodine, Gram Solution
Shelf Life 12 months
CAS 7553-56-2

Specifications

Chemical Name or Material Iodine, Gram Solution
CAS 7553-56-2
Accessories
 

12 months