Please login to your online account to display your discounted pricing

Iodine, Gram Solution

$17.55 - $17.55

Chemical Identifiers

Chemical Name or Material Iodine, Gram Solution
CAS 7553-56-2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S71310

 
fisher scientific
IX1802
0.25N Aqueous Solution 100mL Each for $17.55
Description & Specifications

Specifications

Quantity 100mL
Color Amber
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Packaging 0.25N Aqueous Solution
ChemAlert Storage Symbol Gray
Chemical Name or Material Iodine, Gram Solution
Shelf Life 12 months
CAS 7553-56-2

For staining bacteria

Safety and Handling
 

12 months