Please login to your online account to display your discounted pricing

Iodine, D'Antoni, Certified, 1.5% (w/v) ±0.1%, LabChem™

$114.91 - $114.91

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 95.5
CAS 7681-11-0
CAS Max % 3
CAS 7553-56-2
CAS Max % 1.5
Name Note D'Antoni
Purity Grade Certified Grade
Linear Formula I2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156742

 
lab chem
LC156742
Amber Glass 1L Each for $114.91
Description & Specifications

Specifications

Formula Weight 253.809
Purity Grade Certified Grade
Assay 1.5% ± 0.1% (w/v)
CAS Max % 1.5
Name Note D'Antoni
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 3
Solubility Information Soluble in water
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 7681-11-0
Linear Formula I2
Health Hazard 2 GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life.
Packaging Amber Glass
Identification Passes Test
Physical Form Liquid
CAS 7553-56-2
Quantity 1L
Color Dark red-brown
Chemical Name or Material Iodine
CAS Max % 95.5
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life.