Please login to your online account to display your discounted pricing

Iodine, Certified, 2.5%, 0.200N ±0.002N (0.1M), LabChem™

$335.14 - $335.14

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 92.36
CAS 7681-11-0
CAS Max % 5.08
CAS 7553-56-2
CAS Max % 2.54
CAS 7647-01-0
CAS Max % 0.02
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156624

 
lab chem
LC156624
Amber Glass 4L Each for $335.14
Description & Specifications

Specifications

Formula Weight 253.809
Purity Grade Certified Grade
Assay 2.5%
Identification Passes Test
Health Hazard 1 Warning
Quantity 4L
Color Dark red-brown
CAS 7553-56-2
Packaging Amber Glass
Concentration 0.200N ±0.002N (0.1M)
Chemical Name or Material Iodine
CAS Max % 2.54
Storage Information Room Temperature
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.02
CAS Max % 5.08
CAS 7647-01-0
CAS Max % 92.36
Physical Form Liquid
CAS 7681-11-0
Linear Formula I2
Solubility Information Soluble in water
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Take off contaminated clothing and wash contaminated clothing before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.