Please login to your online account to display your discounted pricing

Iodine, Certified, 12.7%, 1.000N ±0.005N (0.5M), LabChem™

$136.80 - $731.37

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 61.91
CAS 7681-11-0
CAS Max % 25.38
CAS 7553-56-2
CAS Max % 12.69
CAS 7647-01-0
CAS Max % 0.02
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156611

 
lab chem
LC156611
Amber Glass 500mL Each for $136.80

LC156612

 
lab chem
LC156612
Amber Glass 1L Each for $221.29

LC156614

 
lab chem
LC156614
Amber Glass 4L Each for $731.37
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 253.809
CAS 7732-18-5
CAS 7647-01-0
CAS Max % 0.02
Storage Information Room Temperature
CAS 7681-11-0
CAS 7553-56-2
CAS Max % 61.91
Concentration 1.000N ±0.005N (0.5M)
Identification Passes Test
Linear Formula I2
Color Dark red-brown
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.
Physical Form Liquid
Assay 12.7%
Packaging Amber Glass
Quantity 500mL
CAS Max % 25.38
Solubility Information Soluble in water
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 12.69
Chemical Name or Material Iodine
Safety and Handling

Warning

GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash skin with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life.