Please login to your online account to display your discounted pricing

Iodine, Certified, 0.0167N ±0.0001N (0.0084M), LabChem™

$72.29 - $153.66

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 99.35
CAS 7681-11-0
CAS Max % 0.42
CAS 7553-56-2
CAS Max % 0.21
CAS 7647-01-0
CAS Max % 0.02
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156052

 
lab chem
LC156052
Amber Glass 1L Each for $72.29

LC156054

 
lab chem
LC156054
Amber Glass 4L Each for $153.66
Description & Specifications

Specifications

Formula Weight 253.809
Purity Grade Certified Grade
Concentration 0.0167N ±0.0001N (0.0084M)
CAS 7681-11-0
CAS 7732-18-5
Health Hazard 2 GHS H Statement
Harmful to aquatic life.
CAS Max % 99.35
Quantity 1L
CAS 7553-56-2
Identification Passes Test
CAS 7647-01-0
Physical Form Liquid
Linear Formula I2
Storage Information Room Temperature
Chemical Name or Material Iodine
Solubility Information Soluble in water
Packaging Amber Glass
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.42
CAS Max % 0.21
CAS Max % 0.02
Color Red-brown
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life.