missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodine, Certified, 0.0167N ±0.0001N (0.0084M), LabChem™

Manufacturer:  LabChem LC156054

 View more versions of this product

Catalog No. LC156054



Specifications

Specifications

Iodine
99.35,0.42,0.21,0.02
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Soluble in water
Liquid
0.0167N ±0.0001N (0.0084M)
Amber Glass
Red through Brown
7732-18-5,7681-11-0,7553-56-2,7647-01-0
I2
0.0167N ±0.0001N (0.0084M)
253.809
Certified
Passes Test
Room Temperature
4L

Chemical Identifiers

7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions