Please login to your online account to display your discounted pricing

Iodine, Certified, 0.0156N ±0.0001N (0.0078M), LabChem™

$72.29 - $148.83

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7732-18-5
CAS Max % 99.39
CAS 7681-11-0
CAS Max % 0.39
CAS 7553-56-2
CAS Max % 0.2
CAS 7647-01-0
CAS Max % 0.02
Purity Grade Certified Grade
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC156022

 
lab chem
LC156022
Amber Glass 1L Each for $72.29

LC156024

 
lab chem
LC156024
Amber Glass 4L Each for $148.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 253.809
CAS 7553-56-2
Physical Form Liquid
CAS 7647-01-0
Linear Formula I2
Quantity 1L
CAS 7732-18-5
CAS 7681-11-0
Chemical Name or Material Iodine
Concentration 0.0156N ±0.0001N (0.0078M)
Storage Information Room Temperature
CAS Max % 0.02
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.39
Color Red-brown
CAS Max % 0.2
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
Packaging Amber Glass
CAS Max % 99.39
Solubility Information Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.