Please login to your online account to display your discounted pricing

Iodine, ACS Grade, ≥ 99.8%, LabChem™

$836.83 - $836.83

Chemical Identifiers

Chemical Name or Material Iodine
CAS 7553-56-2
CAS Max % 100
Purity Grade ACS Grade
Linear Formula I2
Formula Weight 253.809
UN Number UN3495
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC155901

 
lab chem
LC155901
Amber Glass 500g Each for $836.83
Description & Specifications

Specifications

Assay ≥99.8%
Formula Weight 253.809
Chemical Name or Material Iodine
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Crystals
Linear Formula I2
Storage Information Room Temperature
CAS 7553-56-2
UN Number UN3495
Density 4.93g/cm3
Solubility Information Soluble in water
Health Hazard 1 Danger
Boiling Point 184°C
Melting Point 113°C
Color Purplish-brown
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement
Do not breathe dust, vapors.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Thermal decomposition generates: Iodine vapor
Specific Gravity 4.93g/cm3
Identification Passes Test
Safety and Handling

Danger

GHS P Statement
Do not breathe dust, vapors.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
Very toxic to aquatic life.