missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Iodine, Reagent Grade, ≥99.8% (titration), Honeywell™

Reagent Grade,≥99.8% (titration)

$206.23 - $206.23

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.809
MDL Number MFCD00011007
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60048040 View Documents Honeywell™
I3380100G
100g Glass Bottle Each for $206.23
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7553-56-2
253.809
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
molecular iodine
I2
MFCD00011007
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II

Specifications

7553-56-2
I2
MFCD00011007
3587194
PNDPGZBMCMUPRI-UHFFFAOYSA-N
molecular iodine
807
253.81
Reagent
Glass Bottle
184°C (literature)
100g
100.0000%
Fe2(SO4)3 · xH2O
UN3495
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
253.809
CHEBI:17606
>99.8%
4.930 g/cm3
9 (vs air)
113°C (literature)
Iodine, Reagent Grade, ≥99.8% (titration)
Safety and Handling

Safety and Handling

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Iodine

EINECSNumber : 231-442-4

RTECSNumber : NN1575000

Recommended Storage : Room Temp