Please login to your online account to display your discounted pricing

Iodine, 1.0N Standardized Solution, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Iodine
Physical Form Liquid
Molecular Formula I2
Formula Weight 253.81
MDL Number MFCD00011355
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35632AP

  SDS
alfa aesar
35632AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35632K2

  SDS
alfa aesar
35632K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.81
Quantity 500mL
Chemical Name or Material Iodine
Physical Form Liquid
Molecular Formula I2
MDL Number MFCD00011355