missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodide Standard Solution, 0.100M, Fisher Chemical

$55.82 - $72.18

Chemical Identifiers

CAS 7732-18-5,7681-11-0
CAS Min % 96.0,1.6
CAS Max % 100.0,1.67
Chemical Name or Material Iodide Standard Solution, 0.100M
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SI9001-1 Ricca Chemical Company
RISI90011
Amber Glass 1L Each for $72.18
SI9001-250 SDS Ricca Chemical Company
RISI9001250
Amber Glass 250mL Each for $55.82
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-11-0
96.0,1.6
100.0,1.67
Iodide Standard Solution, 0.100M

Specifications

1L
1.67,100.0
7681-11-0,7732-18-5
Iodide Standard Solution, 0.100M
For AOAC Method 992.22
96.0,1.6
Amber Glass

Safety and Handling