missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iodic Acid, ACS, 99.5%, Alfa Aesar

Manufacturer:  Alfa Aesar 8768118

 View more versions of this product

Catalog No. AA8768118



Specifications

Specifications

Iodic Acid
7782-68-5
99.5%
MFCD00011348
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
OI(=O)=O
175.909
CHEBI:24857
Powder/Chunks
50g
Powder/chunks
HIO3
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
4.629g/cm3
iodic acid
24345
175.91
ACS

Chemical Identifiers

7782-68-5
175.909
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
24345
iodic acid
HIO3
MFCD00011348
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
CHEBI:24857
OI(=O)=O
Safety and Handling

Safety and Handling

H272-H314-H318: May intensify fire; oxidizer. Causes severe skin burns and eye damage. Causes serious eye damage.

EINECSNumber : 231-962-1

SDS Documents