missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iodic acid, 99%

Manufacturer:  Alfa Aesar™ A1192530

 View more versions of this product

Catalog No. AAA1192530


 

Description

Description

Applications
Iodic acid is used in analytical chemistry laboratories to standardize solutions of both weak and strong bases using methyl red or methyl orange as the indicator. It acts as a key starting material to synthesize sodium or potassium iodate, thereby increasing the iodine content of salt.

Solubility
Soluble in water and methanol.

Notes
Light sensitive. Incompatible with reducing agents, alcohols and organic materials.
Specifications

Specifications

Iodic acid
7782-68-5
HIO3
UN3085
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
iodic acid
24345
175.91
110°C (decomposition)
250g
99%
MFCD00011348
14,5012
OI(=O)=O
175.909
CHEBI:24857
Odorless

Chemical Identifiers

7782-68-5
175.909
ICIWUVCWSCSTAQ-UHFFFAOYSA-N
CHEBI:24857
HIO3
MFCD00011348
24345
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents