Invitrogen™ Zero Blunt™ PCR Cloning Kit DFS Item
Offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proofreading, thermostable DNA polymerases
Manufacturer:  Invitrogen™ LSK270020
 View more versions of this product
Catalog No. K270020
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, PCR Cloning
Zero Blunt PCR cloning kits contain linearized pCR-Blunt vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid
20 reactions
TOP10
Cloning, PCR Cloning