Please login to your online account to display your discounted pricing

Invitrogen™ UltraPure™ Urea DFS Item

Effective denaturing agent

$28.67 - $77.25

Specifications

Storage Requirements Store at room temperature.
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

15505050

DFS Item
 
life technologies
LS15505050
2kg Each for $77.25

15505035

DFS Item
 
life technologies
LS15505035
500g Each for $28.67
Description & Specifications

Specifications

Storage Requirements Store at room temperature.

UltraPure Urea is an effective protein denaturant. In a highly concentrated (6 to 8M) urea solution, many proteins unfold and separate into their constituent polypeptide chains. UltraPure Urea is also used to denature nucleic acids in sequencing gels.

Acrylamide Gel Electrophoresis, DNA and RNA Purification and Analysis, Gel Casting, Nucleic Acid Gel Electrophoresis and Blotting, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Shipping Condition: Room temperature