missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ UltraPure™ SSC, 20X
For use in nucleic acid hybridizations and blot transfer applications
Supplier: Invitrogen™ 15557036
Description
UltraPure 20X SSC is a solution formulated and used in concentrations ranging from 0.2X to 20X, depending on the application.
- Supplied in 1L plastic bottles/4L/10L stackable Cubitainer™ Box
- Sterile-filtered UltraPure 20X SSC contains 3.0 M NaCl and 0.3 M sodium citrate, at pH 7.0
- Performance and Quality Testing: No DNase or RNase activity detected
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Specifications
Store at room temperature. | |
SSC | |
Nucleic Acid Gel Electrophoresis, Blotting | |
4 L |
20 X | |
Liquid | |
UltraPure™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.