Please login to your online account to display your discounted pricing
Invitrogen™ TOPO™ XL PCR Cloning Kit, with One Shot™ TOP10 Electrocomp™ E. coli DFS Item
Combines a unique vector and novel protocol for easy, efficient cloning of long (3-10kb) PCR products
Manufacturer: invitrogen™  LSK470010
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. K470010
  • / Each
Description & Specifications
Specifications
For Use With (Application) Cloning, PCR Cloning
Product Type Kit, PCR Cloning
Quantity 10 reactions
Bacterial/Yeast Strain TOP10
Includes Each TOPO XL PCR Cloning Kit includes linearized and topoisomerase I-activated pCR-XL-TOPO vector, dNTPs, control template and primers, M13 forward and reverse primers, sodium iodide solution, binding buffer, 4X final wash, S.N.A.P. purification columns and collection vials, 6X crystal violet loading buffer, 2mg/mL crystal violet solution, One Shot Electrocomp or Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid