Please login to your online account to display your discounted pricing
Invitrogen™ TOPO™ XL PCR Cloning Kit, with One Shot™ TOP10 Electrocomp™ E. coli DFS Item
Combines a unique vector and novel protocol for easy, efficient cloning of long (3-10kb) PCR products
$650.00 - $1,160.00
Specifications
Product Type Kit, PCR Cloning
For Use With (Application) Cloning, PCR Cloning
Includes Each TOPO XL PCR Cloning Kit includes linearized and topoisomerase I-activated pCR-XL-TOPO vector, dNTPs, control template and primers, M13 forward and reverse primers, sodium iodide solution, binding buffer, 4X final wash, S.N.A.P. purification columns and collection vials, 6X crystal violet loading buffer, 2mg/mL crystal violet solution, One Shot Electrocomp or Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid
Quantity 10 reactions
View More Specs
Includes:  Each TOPO™ XL PCR Cloning Kit includes: linearized and topoisomerase I-activated pCR™-XL-TOPO™ vector, dNTPs, control template and primers, M13 forward and reverse primers, sodium iodide solution, binding buffer, 4X final wash, S.N.A.P.™ purification columns and collection vials, 6X crystal violet loading buffer, 2mg/mL crystal violet solution, One Shot™ Electrocomp™ or Chemically Competent E. coli, S.O.C .medium, and pUC19 supercoiled control plasmid.
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Products
Catalog Number Mfr. No. Quantity Price Quantity    
K470010
DFS Item
invitrogen™
LSK470010
10 reactions Each for $650.00
K470020
DFS Item
invitrogen™
LSK470020
20 reactions Each for $1,160.00
Description & Specifications
Specifications
Product Type Kit, PCR Cloning
For Use With (Application) Cloning, PCR Cloning
Includes Each TOPO XL PCR Cloning Kit includes linearized and topoisomerase I-activated pCR-XL-TOPO vector, dNTPs, control template and primers, M13 forward and reverse primers, sodium iodide solution, binding buffer, 4X final wash, S.N.A.P. purification columns and collection vials, 6X crystal violet loading buffer, 2mg/mL crystal violet solution, One Shot Electrocomp or Chemically Competent E. coli, S.O.C. medium, and pUC19 supercoiled control plasmid
Quantity 10 reactions
Topoisomerase I activation allows PCR products to be ligated in just 5 minutes on your bench top and typically results in >80% recombinants. The kit uses the linearized and topoisomerase I-activated pCR™-XL-TOPO vector.
Novel Protocol
The TOPO XL PCR Cloning Kit protocol includes S.N.A.P.™ gel purification which employs crystal violet for improved efficiency of long PCR cloning. This eliminates the need for ethidium bromide and exposure to UV light, which can damage long PCR products and reduce the number of recombinants.
  • ccdB gene for positive selection
  • EcoR I and Nsi I sites flanking the PCR product insertion site for easy excision of inserts
  • Kanamycin and Zeocin™ resistance genes for your choice of selection in E. coli
  • T7 promoter/priming site for in vitro RNA transcription and sequencing
  • M13 forward and reverse primer sites for sequencing
Cloning, PCR Cloning