Please login to your online account to display your discounted pricing
Invitrogen™ TOPO™ TA Cloning™ Kit for Subcloning, with TOP10 E. coli, without manual DFS Item
Provides a highly efficient, 5-minute, one-step cloning strategy (“TOPO Cloning”) for the direct insertion of Taq polymerase–amplified PCR products into a plasmid vector for subcloning
Manufacturer: invitrogen™  LSKNM450040
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. KNM450040
  • / Each
Description & Specifications
Specifications
Bacterial/Yeast Strain TOP10
Includes Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
Quantity 50 reactions
Product Type Kit, Cloning for Subcloning
For Use With (Application) Cloning, PCR Cloning