Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ TOP10F' Chemically Competent E. coli, no manual DFS Item
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
Manufacturer:  Invitrogen™ LSKNM455001
 View more versions of this product
Catalog No. KNM455001
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
TOP10F'
Cloning, PCR Cloning
Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
25 reactions
Kit, Cloning