Please login to your online account to display your discounted pricing
Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ TOP10F' Chemically Competent E. coli, no manual DFS Item
Designed for cloning PCR products directly from a PCR reaction in just 5 minutes
Manufacturer: invitrogen™  LSKNM455040
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. KNM455040
  • / Each
Description & Specifications
Specifications
Product Type Kit, Cloning
Bacterial/Yeast Strain TOP10F'
Includes Box 1: Topoisomerase I-activated pCR2.1-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
For Use With (Application) Cloning, PCR Cloning
Quantity 50 reactions