Please login to your online account to display your discounted pricing
Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP 10 Chemically Competent E.coli, no manual DFS Item
For fast, efficient cloning and subsequent in vitro transcription/translation, use Invitrogen's TOPO TA Cloning Dual Promoter Kit with pCR™II vector
Manufacturer: invitrogen™  LSKNM460040
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. KNM460040
  • / Each
Description & Specifications
Specifications
Product Type Kit, Cloning
Bacterial/Yeast Strain TOP10
Includes Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
For Use With (Application) Cloning, PCR Cloning
Quantity 50 reactions