Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP 10 Chemically Competent E.coli, no manual DFS Item
For fast, efficient cloning and subsequent in vitro transcription/translation, use Invitrogen's TOPO TA Cloning Dual Promoter Kit with pCR™II vector
Manufacturer:  Invitrogen™ LSKNM460001
 View more versions of this product
Catalog No. KNM460001
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, Cloning
Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
25 reactions
TOP10
Cloning, PCR Cloning