Invitrogen™ TOPO™ TA Cloning™ Kit, Dual Promoter, with One Shot™ TOP 10 Chemically Competent E.coli, no manual DFS Item
For fast, efficient cloning and subsequent in vitro transcription/translation, use Invitrogen's TOPO TA Cloning Dual Promoter Kit with pCR™II vector
$816.90 - $1,402.80
Specifications
Product Type Kit, Cloning
For Use With (Application) Cloning, PCR Cloning
Includes Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
Quantity 25 reactions
View More Specs
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Includes: Topoisomerase I-activated pCR™II-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
KNM460040 Invitrogen™
LSKNM460040
50 reactions Each for $1,402.80
KNM460001 Invitrogen™
LSKNM460001
25 reactions Each for $816.90
Specifications
Description
By eliminating time-consuming and tedious restriction site cloning, TOPO cloning is the most reliable method, taking only 5 minutes using a 3-step protocol and yielding up to 95% recombinants.
  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 and SP6 promoters for efficient in vitro transcription
  • M13 forward and reverse primer sites for sequencing
  • 16 convenient and validated restriction sites including EcoR I flanking your insert for subsequent excision or sub-cloning
  • Kanamycin and ampicillin resistance for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants
  • Includes OneShot™ TOP10 F' Competent cells for added convenience and highest cloning efficiency
Cloning, PCR Cloning
Description & Specifications
Specifications
Kit, Cloning
Cloning, PCR Cloning
Box 1: Topoisomerase I-activated pCRII-TOPO vector, PCR buffer, Salt solution, dNTPs, Control template, M13 forward and reverse primers, Control PCR primers, Sterile water. Box 2: One Shot Chemically Competent or Electrocomp E. coli, S.O.C. medium, Supercoiled pUC19 control plasmid
25 reactions