Please login to your online account to display your discounted pricing

ThermoScript™ Reverse Transcriptase DFS Item


An avian RT with reduced RNase H activity designed for RT-PCR

Manufacturer: life technologies  LS12236022

 View more versions of this product

 Disclaimers

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Catalog No. 12236022

  • / Each
 

Description & Specifications

Specifications

Concentration 15U/μL
Temperature (Metric) Operating 65°C
Storage Requirements Store at -70deg.C
Quantity 100 reactions (1500 units)
Test Sensitivity Medium
Packaging Type Tube
Temperature (English) Operating 149°F
Includes 5X cDNA synthesis buffer (250mM Tris acetate (pH 8.4), 375mM potassium acetate, 40mM magnesium acetate, and stabilizers), and 100mM DTT
Format Frozen