Please login to your online account to display your discounted pricing
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli, no manual DFS Item
Designed for cloning Taq-amplified PCR products directly from PCR reaction using overnight ligation step
Manufacturer: invitrogen™  LSKNM204040
 View more versions of this product
 Disclaimers
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. KNM204040
  • / Each
Description & Specifications
Specifications
Vector TA Cloning Vectors
Bacterial Content INVαF'
Includes Linearized pCR2.1vector, T4 DNA ligase, 10X ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls, One Shot TOP10 Chemically Competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Without manual.
Product Type One Shot TOP10 Chemically Competent E. coli
Quantity 40 reactions
Product Name One Shot TOP10 Chemically Competent E. coli
Storage Requirements Store One Shot E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Quantity 40 reactions