Please login to your online account to display your discounted pricing
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli DFS Item
Manufacturer: invitrogen™  LSK204040
 View more versions of this product
Catalog No. K204040
  • / Each
Description & Specifications
Specifications
Includes Linearized pCR2.1 vector, ExpressLink T4 DNA ligase, 5X ExpressLink T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, controls, One Shot TOP10 chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
Product Type One Shot™ TOP10 chemically competent E. coli
Quantity 40 reactions
Bacterial Content TOP10F'
Storage Requirements Store One Shot E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Vector TA Cloning Vectors
Quantity 40 reactions
Product Name One Shot™ TOP10 chemically competent E. coli