Invitrogen™ T4 DNA Ligase (1U/μL) DFS Item
Ideal for cloning (blunt-end or cohesive-end ligation) and adding linkers or adapters to blunt-ended DNA
Manufacturer:  Invitrogen™ LS15224017
 View more versions of this product
Catalog No. 15224017
Specifications
Specifications
1U/μL
5X Reaction Buffer (250mM Tris-HCl (pH 7.6), 50mM MgCl2, 5mM ATP, 5mM DTT, 25% (w/v) polyethylene glycol-8000)
100U
Store at -20°C
Documents
For Research Use Only. Not for use in diagnostic procedures.