missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ SuperScript™ VILO™ cDNA Synthesis Kit DFS Item

Provides the high-temperature capability of SuperScript III Reverse Transcriptase (RT) in an optimized format for generating first-strand cDNA for use in real-time quantitative RT-PCR (qRT-PCR)

Manufacturer:  Invitrogen™ 11754250

 View more versions of this product

Catalog No. 11754250



Description

Description

VILO is an acronym for variable input, linear output; and the SuperScript VILO cDNA Synthesis Kit provides the most consistent results for RT experiments, with unprecedented linearity across the broadest range of input material.

  • Using the SuperScript VILO cDNA Synthesis Kit This formulation provides enhanced cDNA synthesis efficiency and can be used with very low and very high amounts of input RNA (up to 2.5μg total RNA in a 20μL reaction), giving a linear response in message abundance as measured by qPCR. The 10X SuperScript Enzyme Mix includes SuperScript III RT, RNaseOUT Recombinant Ribonuclease Inhibitor, and a proprietary helper protein
  • SuperScript III RT SuperScript III RT is an engineered version of M-MLV RT with reduced RNase H activity and increased thermal stability. The enzyme can be used to synthesize cDNA at a temperature range of 42° to 60°C. Because SuperScript III RT is not significantly inhibited by ribosomal and transfer RNA, it can be used to synthesize cDNA from total RNA. RNaseOUT Recombinant Ribonuclease Inhibitor safeguards against the degradation of target RNA due to ribonuclease contamination
  • Optimized buffer The 5X VILO Reaction Mix includes random primers, MgCl2, and dNTPs in a buffer formulation that has been optimized for qRT-PCR

  • Optimal reaction temperature: 42°C

PCR & Real-Time PCR, Reverse Transcription

Specifications

Specifications

Real-Time Quantitative PCR (qPCR)
42° C
SuperScript™, VILO™
250 reactions
Reduced
250 Reactions
Primer-Probe
Tube
250 Reactions
SuperMix or Master Mix
10 kb or less
250 Reactions
Random Primers
cDNA Synthesis Kit
SuperScript™ III
Dry Ice
10 X
10 X
High
qPCR, RT-PCR
RNA
20 μL
SDS
Documents
Product Certifications
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.