Please login to your online account to display your discounted pricing

Invitrogen™ Qubit™ 2.0 Fluorometer USB DFS Item

For use with the Qubit 2.0 Fluorometer

$67.20 - $115.50

Specifications

For Use With (Equipment) Qubit 2.0 Fluorometer
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Type Price Quantity    

Q32867

DFS Item
 
invitrogen™
LSQ32867
USB Drive Each for $67.20

Q32868

DFS Item
 
invitrogen™
LSQ32868
International Power Cord Each for $115.50
Description & Specifications

Specifications

For Use With (Equipment) Qubit 2.0 Fluorometer

  • Replacement USB drive is intended to be used to transfer data and software updates between the fluorometer and computer
  • Power cord is specifically for use with the Qubit 2.0 Fluorometer and includes four different plug adaptors

Applications:

DNA and RNA Purification and Analysis, DNA Quantitation, Nucleic Acid Quantitation, Protein Quantitation, Proteins, Expression, Isolation and Analysis, RNA Quantitation

Shipping Condition: Room temperature