Please login to your online account to display your discounted pricing
Invitrogen™ Qubit™ 2.0 Fluorometer USB DFS Item
For use with the Qubit 2.0 Fluorometer
Manufacturer: invitrogen™  Q32868
 View more versions of this product
Catalog No. Q32868
  • / Each
Description & Specifications
Specifications
Type International Power Cord
For Use With (Equipment) Qubit 2.0 Fluorometer
  • Replacement USB drive is intended to be used to transfer data and software updates between the fluorometer and computer
  • Power cord is specifically for use with the Qubit 2.0 Fluorometer and includes four different plug adaptors
Applications:
DNA and RNA Purification and Analysis, DNA Quantitation, Nucleic Acid Quantitation, Protein Quantitation, Proteins, Expression, Isolation and Analysis, RNA Quantitation