Invitrogen™ Qubit™ dsDNA HS Assay Kit DFS Item
Highly selective for double-stranded DNA (dsDNA) over RNA
$146.00 - $427.00
Specifications
Requires: Qubit 1.0 or 2.0 Fluorometer or other fluorometer and 500μL thin-walled PCR tubes
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
Q32851 Greener Choice
View Documents
Invitrogen™
LSQ32851
100 assays
Each for $146.00
Q32854 Greener Choice
View Documents
Invitrogen™
LSQ32854
500 assays
Each for $427.00
Description
Description
The Qubit dsDNA HS (High Sensitivity) Assay Kit is designed specifically for use with the Qubit Fluorometer. The assay is highly selective for double-stranded DNA (dsDNA) over RNA and is designed to be accurate for initial sample concentrations from 10 pg/µL to 100 ng/µL. The kit provides concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 µl and 20 µl is acceptable), and read the concentration using the Qubit Fluorometer. Common contaminants, such as salts, free nucleotides, solvents, detergents, or protein are well tolerated in the assay.
Which product to choose for dsDNA HS (High Sensitivity) quantitation?
  • For 1–20 samples: use this Qubit dsDNA HS Assay Kit with the Qubit Fluorometer
  • For 20–2000 samples: use the Quant-iT dsDNA HS Assay Kit with microplate reader
Notes:
1. All Qubit assay kits can be used with the Qubit 1.0, Qubit 2.0, Qubit 3, and Qubit 4 fluorometers.
2. 500 µL thin-walled PCR tubes are required but not included.
3. Also available as a “1X” dsDNA assay kit that provides reagent and buffer in a formulation that is stable as a ready-to-use solution for up to one year after preparation.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
0.0
icon-pdf.png
SDS
For Research Use Only. Not for use in diagnostic procedures.