missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Quant-iT™ PicoGreen™ dsDNA Reagent DFS Item

Selectively detect 25pg/mL of dsDNA in presence of ssDNA, RNA, and free nucleotides

$570.00 - $700.00

Specifications

For Use With (Equipment) Microplate Reader
Range 50pg to 2μg
Sample Type dsDNA
Storage Requirements Store Component A at 2° to 6°C, protect from light.
View More Specs

Products 2
Catalog Number Mfr. No. Includes Quantity Price Quantity  
Catalog Number Mfr. No. Includes Quantity Price Quantity  
P11495 View Documents Invitrogen™
LSP11495
10 vials of 100μL each of Quant-iT PicoGreen dsDNA reagent (Component A). Reagent is sufficient for 200 assays using an assay volume of 2mL. 10 x 100μL Each for $700.00
P7581 View Documents Invitrogen™
LSP7581
1mL in 1 vial of Quant-iT PicoGreen dsDNA reagent (Component A). Reagent is sufficient for 200 assays using an assay volume of 2mL. 1mL Each for $570.00
Description

Description

PicoGreen dsDNA quantitation assay is linear over three orders of magnitude and has little sequence dependence, allowing accurate measurement of DNA from many sources, including genomic DNA, viral DNA, miniprep DNA, or PCR amplification products.

  • Orders of magnitude more sensitive than UV absorbance readings, saving on precious sample
  • Specific for dsDNA in presence of equimolar amounts of RNA
  • Easy to use-just add dye to sample, wait 5 minutes and read
  • Suitable for use in 96- and 384-well plate formats
  • Compatible with most fluorescence-based microplate readers and fluorometers and fluorescence
  • PCR-based assays, microarray samples, DNA damage assays, enzyme activity assays, genomic DNA quantitation, measuring dsDNA in complex mixtures and viral DNA quantitation

DNA and RNA Purification and Analysis, DNA Quantitation, Nucleic Acid Quantitation

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Microplate Reader
50pg to 2μg
dsDNA
Store Component A at 2° to 6°C, protect from light.

For Research Use Only. Not for use in diagnostic procedures.