Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ Quick Gel Extraction Kit DFS Item


Purification protocols are provided for centrifugation and vacuum

Manufacturer: invitrogen™  LSK210012

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K210012

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing
Isolation Technology Silica Spin Column, Spin or Vacuum Column
Target Nucleic Acids in Gel Fragments
Sample Type DNA Fragments in Agarose Gel Slices
Quantity 50 preps
Quantity 50 preps