Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ PCR Purification Kit DFS Item


No tedious gel purification necessary

Manufacturer: invitrogen™  LSK310002

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K310002

  • / Each
 

Description & Specifications

Specifications

Includes
  • 72mL Binding Buffer (B2)
  • 109mL Binding Buffer High-Cutoff (B3)
  • 80mL Wash Buffer (W1)
  • 15mL Elution Buffer (E1)
  • 5 × 50 PureLink™ PCR Spin Columns with Collection Tubes
  • 5 × 50 PureLink™ Elution Tubes (1.7mL)
Storage Requirements Store at room temperature.
Isolation Technology Silica Spin Column
Quantity 250 preps
Target DNA from Enzymatic Reactions (e.g. PCR)
Sample Type PCR Products
For Use With (Application) Cloning, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing
Quantity 250 preps