Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ PCR Micro Kit DFS Item


Silica Spin Column

Manufacturer: invitrogen™  LSK310010

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K310010

  • / Each
 

Description & Specifications

Specifications

Includes Columns, binding and wash buffers, recovery tubes, and the manual
Separation Range 100bp to 12kb
Volume (Metric) Sample 125μL
Isolation Technology Silica Spin Column
Yield 5 - 20μL
Test Time 10 min.
For Use With (Application) Cloning, Sequencing, Nucleic Acid Labeling, PCR
Sample Type Enzymatic Reactions (DNA (Restriction Enzyme-Digested), PCR Products, cDNA Labeling Reactions)
Storage Requirements Room Temperature
Quantity 10 columns/preps