Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ HiPure Plasmid FP (Filter and Precipitator) Maxiprep Kit DFS Item


From cell pellet to ultrapure DNA in less than one hour

Manufacturer: invitrogen™  LSK210026

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K210026

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Sample Size > 200 μg (Large-Scale) Plasmid DNA
Yield 1000 μg
Isolation Technology Anion Exchange Resin with Lysate Clearing Filter & Precipitator
Target Plasmid DNA
Sample Type Bacterial Culture
Quantity 10 preps
Quantity 10 preps