missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ HiPure Plasmid FP (Filter and Precipitator) Maxiprep Kit DFS Item

From cell pellet to ultrapure DNA in less than one hour

$456.00 - $1,088.00

Specifications

For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Isolation Technology Anion Exchange Resin with Lysate Clearing Filter & Precipitator
Sample Size > 200 μg (Large-Scale) Plasmid DNA
Sample Type Bacterial Culture
Target Plasmid DNA
View More Specs

Includes: The PureLink HiPure Plasmid FP Maxiprep Kit includes Equilibration Buffer, Resuspension Buffer, Lysis Buffer, Precipitation Buffer, RNase A, Wash Buffer, Elution Buffer, TE Buffer, HiPure Filter Columns, and a PureLink HiPure Precipitator Module. The PureLink HiPure Precipitator Module includes HiPure Precipitators, and 5mL and 30mL syringes.

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
K210027 View Documents Invitrogen™
LSK210027
25 preps Each for $1,088.00
K210026 View Documents Invitrogen™
LSK210026
10 preps Each for $456.00
Description

Description

Transfection grade. The PureLink HiPure Plasmid FP (Filter and Precipitator) Maxiprep Kit isolates plasmid DNA. Using HiPure Filter Columns and HiPure Precipitators supplied in kit, you can purify plasmid DNA without centrifugation steps required in traditional isolation protocols. The quality of purified plasmid DNA is equivalent to that obtained by two passes through cesium chloride gradients - most rigorous method for DNA purification - making it suitable for transfection and other applications requiring high-purity plasmid DNA.

  • Simple protocols - bacterial lysate clarification and isopropanol precipitation without centrifugation
  • Fast results - from E. coli cell pellet to high-purity, transfection-grade plasmid DNA in about 60 minutes
  • High yields - up to 850μg from a high-copy plasmid

DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Plasmid DNA Purification

Specifications

Specifications

Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
> 200 μg (Large-Scale) Plasmid DNA
Plasmid DNA
Anion Exchange Resin with Lysate Clearing Filter & Precipitator
Bacterial Culture
1000 μg

For Research Use Only. Not for use in diagnostic procedures.