Please login to your online account to display your discounted pricing

Invitrogen™ PureLink™ HiPure Plasmid Filter Midiprep Kit DFS Item


Purification in 90 minutes

Manufacturer: invitrogen™  LSK210015

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. K210015

  • / Each
 

Description & Specifications

Specifications

Sample Size 100-200 μg (Medium-Scale) Plasmid DNA
Yield 350 μg
Sample Type Bacterial Culture
Isolation Technology Anion Exchange Resin with Lysate Clearing Filter
For Use With (Application) Cloning, In Vitro Transcription, Next-Generation Sequencing, Nucleic Acid Labeling, PCR, Sequencing, Transfection, Transformation
Quantity 50 preps
Target Plasmid DNA
Quantity 50 preps