Invitrogen™ SuperScript™ III Platinum™ SYBR™ Green One-Step qRT-PCR Kit DFS Item
Combine the most powerful reverse transcriptase and DNA polymerase technologies to deliver precise and accurate analysis of gene expression in a convenient one-step format.
Manufacturer:  Invitrogen™ LS11736051
 View more versions of this product
Catalog No. 11736051
Description
Description
  • SuperScript™ III RT for higher temperature cDNA synthesis for greater success with difficult RNA secondary structure
  • Platinum™ Taq DNA Polymerase with hot-start technology for improved specificity
  • LUX™ Fluorogenic Primers (Figure 1) or dual-labeled fluorogenic probes for superior detection performance
  • SYBR™ Green I dye for easy and convenient detection
  • Validated performance on multiple real-time instrument platforms, including rotor-based systems
Order Info
Shipping Conditions: Dry Ice
Specifications
Specifications
• 100 µl SuperScript® III RT/Platinum® Taq Mix
• 2 × 1.25 ml 2X SYBR® Green Reaction Mix
• 1 ml Magnesium Sulfate (50 mM)
• 100 µl ROX Reference Dye (25 µM)
• 300 µl 20X Bovine Serum Albumin (1 mg/ml)
Store all components at -20°C (-80°C for long-term storage). ROX Reference Dye must be stored in the dark.
Platinum®, SYBR®, SuperScript™
Dry Ice
1-Step qRT-PCR
100 Reactions
7500 System, BioRad iCycler iQ, BioRad iQ5, Stratagene Mx4000, MJ Chromo4, MJ Opticon, Stratagene Mx3000P, Stratagene Mx3005P, Cepheid SmartCycler, BioRad MyiQ
RNA
SYBR
Standard
SDS
Documents
For Research Use Only. Not for use in diagnostic procedures.